Diode laser, red
Diode laser, red, 650 nm
Katalog # DIDAPOD013135
Leverandør: DIDALAB
Gå til:
Product Details & Documents
The diameter of the beam is adjustable from 1 to 12 mm.
: Supplied with Ø 10-mm rod and mains adapter.
Specifikationer
- Beskrivelse:Diode laser, red
- Bølgelængde:650 nm