Orders LinkContactUsLinkComponent
Sucrose solutions for refractometer calibration
Sucrose solutions for refractometer calibration
1,34782, 10 °Brix , 15 mL
Katalog # BELLSS10
Leverandør:   BELLINGHAM STANLEY
Sucrose solutions for refractometer calibration
Katalog # BELLSS10
Leverandør:  BELLINGHAM STANLEY
CAS nummer:  
Sucrose solutions for refractometer calibration
Sucrose solutions for refractometer calibration
1,34782, 10 °Brix , 15 mL
CAS nummer:  
Price
517,00 kr
Log ind for tilgængelighed
1 * 15 mL
Product Details & Documents
Sucrose solutions are an extremely easy to use medium for verifying and calibrating refractometers measuring in the Brix scale, as no special consideration is required when testing instruments at ambient temperature fitted with ATC.
  • Uncertainty: ±0,02 °Brix
  • Traceable to NIST and ICUMSA
  • Supplied in 15 ml plastic dropper bottles for ease of use
  • Sucrose solutions have a limited shelf life of 6 weeks
Sucrose solutions are manufactured on a %weight/weight basis and are calibrated in accordance with EN ISO/IEC 17025:2005.
Documents
Specifikationer
Specifikationer
  • Pakn:
    15 mL
  • Refraktionsindeks:
    1,34782
  • Specifikation:
    10 °Brix