Orders LinkContactUsLinkComponent
VWR® PCR DNA Marker
VWR® PCR DNA Marker
PCR DNA Marker™, 1 KIT
Katalog # E854-100RXN
Leverandør:   VWR Chemicals
VWR® PCR DNA Marker
Katalog # E854-100RXN
Leverandør:  VWR Chemicals
CAS nummer:  
VWR® PCR DNA Marker
VWR® PCR DNA Marker
PCR DNA Marker™, 1 KIT
CAS nummer:  
Price
2.060,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
PCR DNA Marker™ offers 8 fragments ranging from 50 to 2000 bp. The marker contains high intensity reference bands that are ideal for quick sizing of PCR products and restriction digests. Simply mix 10 μl of PCR DNA Marker™ with required quantity of loading buffer and load into well. There is sufficient material for 100 assays.
  • High resolution marker optimal for size estimation of PCR fragments
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    PCR DNA Marker™