Accessories for Loopster basic and digital
Leverandør: IKA
Gå til:
Om denne vare
11 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
- TilgængelighedPrice
- 444-0850Tube attachment for 48×1.5 ml tubes with Ø10 mm
- 444-0851Tube attachment for 24×5 ml tubes with Ø12 mm
- 444-0852Tube attachment for 20×15 ml tubes with Ø16 mm
- 444-0853Tube attachment for 12×30 ml tubes with Ø25 mm
- 444-0854Tube attachment for 12×50 ml tubes with Ø28 mm
- 444-0855Universal disc, spare clips (available separately) can be arranged as required on the discUniversal disc, spare clips (available separately) can be arranged as required on the discLog ind for tilgængelighed1 * 1 Items1.270,00 kr
- 444-0856Spare clips with Ø10 mm for use with Universal discSpare clips with Ø10 mm for use with Universal discLog ind for tilgængelighed1 * 12 Items384,00 kr
- 444-0857Spare clips with Ø12 mm for use with Universal discSpare clips with Ø12 mm for use with Universal discLog ind for tilgængelighed1 * 12 Items384,00 kr
- 444-0951Spare clips with Ø16 mm for use with Universal discSpare clips with Ø16 mm for use with Universal discLog ind for tilgængelighed1 * 12 Items384,00 kr
- 444-0952Spare clips with Ø25 mm for use with Universal discSpare clips with Ø25 mm for use with Universal discLog ind for tilgængelighed1 * 12 Items384,00 kr
- 444-0953Spare clips with Ø28 mm for use with Universal discSpare clips with Ø28 mm for use with Universal discLog ind for tilgængelighed1 * 12 Items384,00 kr
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Spare clips with Ø25 mm for use with Universal discSpare clips with Ø25 mm for use with Universal disc444-0952Log ind for tilgængelighed1 * 12 Items - 384,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Spare clips with Ø12 mm for use with Universal discSpare clips with Ø12 mm for use with Universal disc444-0857Log ind for tilgængelighed1 * 12 Items - 384,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Spare clips with Ø10 mm for use with Universal discSpare clips with Ø10 mm for use with Universal disc444-0856Log ind for tilgængelighed1 * 12 Items - 384,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Spare clips with Ø28 mm for use with Universal discSpare clips with Ø28 mm for use with Universal disc444-0953Log ind for tilgængelighed1 * 12 Items - 384,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Spare clips with Ø16 mm for use with Universal discSpare clips with Ø16 mm for use with Universal disc444-0951Log ind for tilgængelighed1 * 12 Items - 384,00 kr