4-Bromo-1H-pyrazole
Glass bottle for solids, 100 g
Katalog # ACTIAS20439G100
Marketsource
Leverandør: ACTIVATE SCIENTIFIC
4-Bromo-1H-pyrazole
Katalog # ACTIAS20439G100
Leverandør: ACTIVATE SCIENTIFIC
Marketsource
CAS nummer: 2075-45-8
Gå til:
Product Details & Documents
- CAS nummer:2075-45-8
- Formel:C₃H₃BrN₂
- MDL nummer:MFCD00075602
- Molecular Weight:146.97 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



