Slipper animalcule
Slipper animalcule, SOMSO-Plast®, 2,7 kg, 390×260×610 mm
Katalog # SOMMZOS107
Leverandør: SOMMER MARCUS
Gå til:
Product Details & Documents
Paramecium. The model shows the cell inventory of a portozoa: Macro- and micronucleus, contractile vacuoles, cytostome with membranellae, myonemes and food vacuoles and the formation of the endo- and ectoplasm and the network of neuronemes.
- Model enlarged roughly 1600 times
- Made in SOMSO-Plast®
- Model separates into two parts
A detailed block shows the pellicle of the ectoplasm, the position and order of the trichocysts, and a range of cilia in typical order.
: On a stand with green base and explanatory notes.
Specifikationer
- Vægt:2,7 kg
- Beskrivelse:Slipper animalcule
- Materiale:SOMSO-Plast®
- B×D×H:390×260×610 mm