Orders LinkContactUsLinkComponent
Stomach wall
Stomach wall
Stomach wall, SOMSO-Plast®, 1,3 kg, 300×190×180 mm
Katalog # SOMMJS6
Leverandør:   SOMMER MARCUS
Stomach wall
Katalog # SOMMJS6
Leverandør:  SOMMER MARCUS
Stomach wall
Stomach wall
Stomach wall, SOMSO-Plast®, 1,3 kg, 300×190×180 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The formation and structure of layers are shown by a transverse and vertical section.
  • Enlarged many times
Leveringsinformation: On a green base plate.
Specifikationer
  • Vægt:
    1,3 kg
  • Beskrivelse:
    Stomach wall
  • Materiale:
    SOMSO-Plast®
  • B×D×H:
    300×190×180 mm