Section through the elbow
Section through the elbow
Katalog # SOMMNS46
Leverandør: SOMMER MARCUS
Gå til:
Product Details & Documents
Sagittal section. Model in one piece.
- Natural size
- Made in SOMSO-Plast®
H×W×D: 26×32×4 cm
Weight: 0,8 kg
Specifikationer
- Beskrivelse:Section through the elbow