Accessories for optical bench components
Leverandør: DIDALAB
Gå til:
Om denne vare
6 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
- TilgængelighedPrice
- DIDAPOD010080Lens holder for fragile components, Ø 80 mm
- DIDAPOD010090Lens holder, Ø 40/42 mm
- DIDAPOD060130Lens holder, Ø 80 mm
- DIDAPOD010095Permanent frame, Ø 40 mm
- DIDAPOD010110Simple lens holder, Ø 40 mm
- DIDAPOD002192Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mmStand, half moon, for optical components positioned remote from the Optical bench, height 210 mmLog ind for tilgængelighed1 * 1 Items940,00 kr