Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
732-1475
: Brand
- Optisk egenskab:Transparent
- Beskrivelse:Sealing film, self-adhesive, for ELISA, Deep Well and PCR plates, polyester, single sheets
- Pakn:100
Highly transparent film made of polyester is suitable for Real-Time PCR measurements. Temperature range: −40 to +120 °C.