Orders LinkContactUsLinkComponent
Pipette tips, ClickSure™ Tips
Leverandør:   HAMILTON BONADUZ
Pipette tips, ClickSure™ Tips
Leverandør:  HAMILTON BONADUZ
Certifikater
Pipette tips, ClickSure™ Tips
Pipette tips, ClickSure™ Tips
4 tilgængelige muligheder nedenfor
Product Details & Documents
These tips are validated to optimise the performance of the Microlab 300 system, offering superior accuracy and precision. When the tips are placed onto the probe, the O-ring on the probe compresses into the tip groove. This design only requires a low insertion force to successfully load a tip, resulting in a more ergonomic experience for the user and enabling long-term use with minimal energy exerted.
  • Tip packaging and tips are recyclable for eco-conscious labs
  • Less force is required to load and eject a tip
Product Family Options
Produktinformation
  • Spidstype
    Volumen
    Steril
    Pakket
    Tilgængelighed
    Price
  • Standard
    50 µl
    -
    Bulk, 960/bag
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 664,00 kr
    664,00 kr
    1 * 1 Items
       664,00 kr
  • Standard
    50 µl
    -
    10 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 1.860,00 kr
    1.860,00 kr
    1 * 10 Items
       1.860,00 kr
  • Standard
    1000 µl
    -
    Bulk, 960/bag
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.870,00 kr
    1.870,00 kr
    1 * 1 Items
       1.870,00 kr
  • Standard
    1000 µl
    -
    10 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 2.020,00 kr
    2.020,00 kr
    1 * 10 Items
       2.020,00 kr