UHPLC columns, Pinnacle® DB PFP Propyl (USP L43)
: Restek
Pinnacle® DB PFP Propyl, a unique pentafluorophenyl phase with a propyl spacer, uses a highly base-deactivated spherical silica manufactured by Restek. This highly base-deactivated packing exhibits excellent peak shapes for a wide range of compounds, including nucleosides, nucleotides and halogenated compounds.
- Catalog No:
- REST941950212
- REST9419362
- REST9419525
- REST9419355
- REST9419562
- 554-1569
- 554-1568
- REST9419352
- 554-1567
- REST9419515
- REST9419552
- REST9419512
- REST9419575
- REST9419332
- REST9419365
- REST9419572
- REST9419565
- REST941950210
- Kulstofbelastning:
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- Endcapped:
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
Frequently Bought Together