High visibility winter jackets, Eagle 050A
Supplier: SIOEN
About this item
Multifunctional jacket made of 80% polyester/20% cotton. With fixed polyamide lining in body, quilted sleeves and detachable fur lining with one patched pocket.
- Detachable fur collar and sleeves
- Zip closure, knitted windcuffs at end of sleeves
- Two patched chest pockets with zip closure, two inset pockets with zip closure, one inside pocket in lining
- Elasticated waist
Back length: 75 cm
Weight: 280 g/m²
Certifications: EN 471 (3-2, with sleeves)
6 Options Available Below
Product Details & Documents
Multifunctional jacket made of 80% polyester/20% cotton. With fixed polyamide lining in body, quilted sleeves and detachable fur lining with one patched pocket.
- Detachable fur collar and sleeves
- Zip closure, knitted windcuffs at end of sleeves
- Two patched chest pockets with zip closure, two inset pockets with zip closure, one inside pocket in lining
- Elasticated waist
Back length: 75 cm
Weight: 280 g/m²
Certifications: EN 471 (3-2, with sleeves)
Specifications for Product Family
Specifications
- Catalog No:
- SIOE050A/FC1/S
- SIOE050A/FC1/M
- SIOE050A/FC1/L
- SIOE050A/FC1/1XL
- SIOE050A/FC1/2XL
- SIOE050A/FC1/3XL
- Model:
- 050A Eagle
- 050A Eagle
- 050A Eagle
- 050A Eagle
- 050A Eagle
- 050A Eagle
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold|HiVis
- Cold|HiVis
- Cold|HiVis
- Cold|HiVis
- Cold|HiVis
- Cold|HiVis
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton
- 80% polyester, 20% cotton
- Vægt:
- 280 g/m²
- 280 g/m²
- 280 g/m²
- 280 g/m²
- 280 g/m²
- 280 g/m²
Specifications
Product Family Options
Product Information
- FarveTøj størrelseAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/SModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m² - Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/MModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m² - Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/LModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m² - Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/1XLModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m² - Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/2XLModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m² - Specifications:
More
Specifications
Cat. No.SIOE050A/FC1/3XLModel050A EagleOutfit applikationerGeneral PurposeRisk areaCold|HiVisKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% polyester, 20% cottonVægt280 g/m²