Hood, 446
hvid
Katalog # MMMA446
Leverandør: 3M
Gå til:
Product Details & Documents
This hood is made of the same microporous laminated PP/PE fabric used in the 3M™ overall 4510. The hood offers a general barrier to non hazardous substances, with drapes over the shoulders offering excellent coverage.
- Low-linting fabric
- Anti-static treated
- Serged seams
- Elasticated around the face
Cat. I
: Cat. I
Specifikationer
- Farve:hvid
- Cat. No.:MMMA446
- Model:446
- Outfit applikationer:Laboratory