Orders LinkContactUsLinkComponent
EC medium MPN tubes
EC medium MPN tubes
Supplier:   Hach
EC medium MPN tubes
Supplier:  Hach
Certificates
EC medium MPN tubes
EC medium MPN tubes
2 Options Available Below
Product Details & Documents
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements
Documents
Product Family Options
Product Information
  • Beskrivelse
    Standard
    Pakn
    Availability
    Price
  • EC medium MPN tubes
    -
    15
    Log in for availability
    Log in for availability
    1 * 15 Items
    1 * 15 Items - 529,00 kr
    529,00 kr
    1 * 15 Items
       529,00 kr
  • EC/MUG without durham tubes
    -
    15
    Log in for availability
    Log in for availability
    1 * 15 Items
    1 * 15 Items - 604,00 kr
    604,00 kr
    1 * 15 Items
       604,00 kr