Orders LinkContactUsLinkComponent
Flow-through filters
Flow-through filters
Marketsource
Leverandør:   Roth Carl
Flow-through filters
Leverandør:  Roth Carl
Marketsource
Certifikater
2 tilgængelige muligheder nedenfor
Product Details & Documents
These flow-through filters are made of PTFE. The filter membrane can be replaced easily by unscrewing the sample funnel. Plastic tubing can be connected with olives.
  • Suitable for overpressure or vacuum up to about 5 bar
  • Matching adapter for 2× tube sizes included
  • Anti-adhesive surface allows easy cleaning
Delivery information: Delivery without filter membranes, please order separately.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • ROTHY935.1
  • ROTHY937.1
  • Filtre:
  • Ø 25 mm
  • Ø 90 mm
  • Filtreringsareal:
  • 3,1 cm²
  • 52,0 cm²
  • Max. temperature:
  • 121 °C
  • 121 °C
Specifikationer
Product Family Options
Produktinformation
  • Model
    Ø
    Tilgængelighed
    Price
  • ROTHY935.1
    Flow-through filters
    Flow-through filters
    GL 14
    25 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.531,53 kr
    2.531,53 kr
    1 * 1 Items
       2.531,53 kr
    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ROTHY935.1
    Filtre
    Ø 25 mm
    Filtreringsareal
    3,1 cm²
    Max. temperature
    121 °C
    Marketsource
  • ROTHY937.1
    Flow-through filters
    Flow-through filters
    GL 25
    90 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 6.213,07 kr
    6.213,07 kr
    1 * 1 Items
       6.213,07 kr
    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ROTHY937.1
    Filtre
    Ø 90 mm
    Filtreringsareal
    52,0 cm²
    Max. temperature
    121 °C
    Marketsource