Orders LinkContactUsLinkComponent
Brushes, FlexBrush™
Brushes, FlexBrush™
   Sustainable Options Available
Supplier:   MICRONOVA
Brushes, FlexBrush™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Brushes, FlexBrush™
6 Options Available Below
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications for Product Family
Specifications
  • Catalog No:
  • 129-4202
  • MICRBRWM-22512S_U
  • 129-1068
  • 141-1016
  • 141-1644
  • 141-1645
  • Environmentally Preferable:
  • Reduktion af affald
  • Reduktion af affald
  • Reduktion af affald
  • Reduktion af affald
  • Reduktion af affald
Specifications
Product Family Options
Brushes
Product Information
  • Beskrivelse
    Børstemateriale
    Børstehoved Ø
    Handle material
    Samlet længde
    Availability
    Price
  • J-Brush adapter, hook-shaped brush, stainless steel with plastic handle
    Nylon
    57,1 mm
    Plast
    406 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 624,00 kr
    624,00 kr
    1 * 1 Items
       624,00 kr
    Specifications:
      Reduktion af affald
    More
    Specifications
    Cat. No.
    129-4202
    Environmentally Preferable
    Reduktion af affald
      Reduktion af affald
  • S-Brush adapter, S-shaped brush, stainless steel with plastic handle
    Nylon
    57,1 mm
    Rustfrit stål
    533 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    MICRBRWM-22512S_U
    Environmentally Preferable
  • Adapter, hook-shaped brush, twisted zinc with plastic handle
    Polyester
    Plast
    292 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 437,00 kr
    437,00 kr
    1 * 1 Items
       437,00 kr
    Specifications:
      Reduktion af affald
    More
    Specifications
    Cat. No.
    129-1068
    Environmentally Preferable
    Reduktion af affald
      Reduktion af affald
Covers
Product Information
  • Beskrivelse
    Børstemateriale
    Børstehoved Ø
    Handle material
    Samlet længde
    Availability
    Price
  • Polyester cover, NovaPoly
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 186,00 kr
    186,00 kr
    1 * 1 Items
       186,00 kr
    Specifications:
      Reduktion af affald
    More
    Specifications
    Cat. No.
    141-1016
    Environmentally Preferable
    Reduktion af affald
      Reduktion af affald
  • Polyester cover, irradiated, NovaPoly
    Log in for availability
    Log in for availability
    1 * 12 Items
    1 * 12 Items - 3.010,00 kr
    3.010,00 kr
    1 * 12 Items
       3.010,00 kr
    Specifications:
      Reduktion af affald
    More
    Specifications
    Cat. No.
    141-1644
    Environmentally Preferable
    Reduktion af affald
      Reduktion af affald
  • Microfibre cover (80% Polyester, 20% Polyamide)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 209,00 kr
    209,00 kr
    1 * 1 Items
       209,00 kr
    Specifications:
      Reduktion af affald
    More
    Specifications
    Cat. No.
    141-1645
    Environmentally Preferable
    Reduktion af affald
      Reduktion af affald