Multi-element kvalitetskontrolstandard 10 mg/l 12 elementer i salpetersyre 2% / Flussyre < 0,1% ARISTAR® til ICP-MS
Catalog # 84794.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pakning:Plastic bottle
- Pakn:100 mL
- Beskrivelse:Multi-element kvalitetskontrolstandard 10 mg/l {Hf; Ge; Mo; Nb; Sb; Si; Sn; Ta; Te; Ti; W; Zr} i salpetersyre 2% / Flussyre < 0,1% ARISTAR® til ICP-MS
Specifications
About this item
12 elements at 10 mg/L: Hf, Ge, Mo, Nb, Sb, Si, Sn, Ta, Te, Ti, W, Zr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.