Orders LinkContactUsLinkComponent
Pipette Tips, Standard
   Bæredygtige muligheder
Leverandør:   Brand
Pipette Tips, Standard
Leverandør:  Brand
   Bæredygtige muligheder
Certifikater
Pipette Tips, Standard
68 tilgængelige muligheder nedenfor
Product Details & Documents
These tips are manufactured under cleanroom conditions and are automatically packaged to ensure that the tips are consistently free from contamination.
  • High-purity PP, free from DiHEMDA and oleamide, manufactured without lubricants; cadmium-free pigments
  • Rack-packed pipette tips up to 1250 µl are DNA, RNases, endotoxins and ATP-free, independent of the packaging
  • Graduation for a quick volume check
  • Sterile tips and packaging are manufactured exclusively under BIO-CERT® LIQUID HANDLING QUALITY certification
  • Autoclavable at 121 °C (2 bar), acc. DIN EN 285
The tips are tested for compatibility with BRAND pipettes and are suitable for use with most of the common pipettor brands including GILSON®, Thermo Fisher Scientific FINNPIPETTE®, Eppendorf® and Sartorius. The 5 ml tip is exclusively tested for BRAND pipettes and Thermo Fisher Scientific FINNPIPETTE®. The 10 ml/1250 µl tip is suitable for BRAND, Eppendorf®.
Bulk packaging in reclosable bags, non sterile
Tips are produced under supervised cleanroom conditions and automatically shrink wrapped in reclosable bags and packaged in cardboard boxes. The batch number is printed on every bag.
TipBox, non sterile and sterile, BIO-CERT® LIQUID HANDLING QUALITY
Tip box for all sizes up to 1250 µl. With transparent window, hinged and push-on lid (dual function closure) and coloured tip trays, with side panel labelling. Usable with a partially loaded multi channel pipette. The 5 ml and 10 ml tips come in a corresponding round tip box for large tips.
TipRack, non sterile and sterile, BIO-CERT® LIQUID HANDLING QUALITY
Space saving refill unit for TipBox, with environmentally compatible packaging made of recyclable PET. Compared to the filled TipBoxes, the amount of waste is reduced by 20%. Sterile racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilised TipBox. All tip-trays are printed on one side with information on the contents.
BIO-CERT® LIQUID HANDLING QUALITY, sterile: These tips are made from high-quality, pure PP without any mould release agents and other additives. All lots are tested for sterility (acc. ISO 11 137 and AAMI guidelines, a SAL of 10⁻⁶ is obtained) and are certified free from endotoxins, DNA-, RNase- and ATP. Tips are supplied with a Certificate of Analysis.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • BRND732182
  • 613-2318
  • BRND732184
  • 613-2326
  • 613-2333
  • BRND732188
  • 613-2428
  • BRND732190
  • 613-2339
  • BRND732192
  • 613-2401
  • 613-2319
  • 613-2327
  • 613-2334
  • 613-2429
  • 613-2340
  • 613-2402
  • 613-2322
  • 613-2328
  • 613-2335
  • 613-2430
  • 613-2341
  • 613-2403
  • 613-2323
  • 613-2329
  • 613-2336
  • 613-2431
  • 613-2342
  • 613-2407
  • 613-2324
  • 613-2331
  • 613-2337
  • 613-2432
  • 613-2343
  • 613-2408
  • 613-2325
  • 613-2332
  • 613-2338
  • 613-2433
  • 613-2344
  • 613-2409
  • 613-7275
  • 613-7277
  • 613-7171
  • 613-7173
  • 613-7175
  • 613-7163
  • 732-3971
  • 732-3973
  • 732-3976
  • 732-3969
  • 732-3975
  • 732-3972
  • 732-3974
  • 732-3970
  • 613-7276
  • 613-7178
  • 613-7177
  • 613-7167
  • 613-7165
  • 613-7172
  • 612-5754
  • 612-5701
  • 613-3841
  • 613-3445
  • 613-3446
  • 613-3842
  • BRND732186
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Reduktion af affald|Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
Specifikationer
Product Family Options
0,1 - 20 µl, length: 37 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked, BIO-CERT® Liquid handling quality
    0,1 - 20 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 619,00 kr
    619,00 kr
    1 * 960 Items
       619,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732182
    Environmentally Preferable
    New Product
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    0,1 - 20 µl
    -
    2 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2.000 Items
    1 * 2.000 Items - 755,00 kr
    755,00 kr
    1 * 2.000 Items
       755,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2318
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk,CERTIFIED LIFE SCIENCE QUALITY
    0,1 - 20 µl
    -
    10 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10.000 Items
    1 * 10.000 Items - 3.030,00 kr
    3.030,00 kr
    1 * 10.000 Items
       3.030,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2326
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    0,1 - 20 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 496,00 kr
    496,00 kr
    1 * 960 Items
       496,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2333
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    0,1 - 20 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 900,00 kr
    900,00 kr
    1 * 960 Items
       900,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2428
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    0,1 - 20 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 433,00 kr
    433,00 kr
    1 * 480 Items
       433,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2339
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    0,1 - 20 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 945,00 kr
    945,00 kr
    1 * 960 Items
       945,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2401
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
0,5 - 20 µl, length: 46 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked, BIO-CERT® Liquid handling quality
    0,5 - 20 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 579,00 kr
    579,00 kr
    1 * 960 Items
       579,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732184
    Environmentally Preferable
    New Product
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    0,5 - 20 µl
    -
    2 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2.000 Items
    1 * 2.000 Items - 704,00 kr
    704,00 kr
    1 * 2.000 Items
       704,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2319
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    0,5 - 20 µl
    -
    10 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10.000 Items
    1 * 10.000 Items - 2.830,00 kr
    2.830,00 kr
    1 * 10.000 Items
       2.830,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2327
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    0,5 - 20 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 463,00 kr
    463,00 kr
    1 * 960 Items
       463,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2334
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    0,5 - 20 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 837,00 kr
    837,00 kr
    1 * 960 Items
       837,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2429
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    0,5 - 20 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 405,00 kr
    405,00 kr
    1 * 480 Items
       405,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2340
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    0,5 - 20 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 881,00 kr
    881,00 kr
    1 * 960 Items
       881,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2402
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
2 - 200 µl, length: 50 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked BIO-CERT® Liquid Handling Quality
    2 - 200 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 536,00 kr
    536,00 kr
    1 * 960 Items
       536,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732188
    Environmentally Preferable
    New Product
  • Bulk, yellow, CERTIFIED LIFE SCIENCE QUALITY
    2 - 200 µl
    -
    1 pose, 1000
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 234,00 kr
    234,00 kr
    1 * 1.000 Items
       234,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2323
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, yellow, CERTIFIED LIFE SCIENCE QUALITY
    2 - 200 µl
    -
    10 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10.000 Items
    1 * 10.000 Items - 1.910,00 kr
    1.910,00 kr
    1 * 10.000 Items
       1.910,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2329
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    2 - 200 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 429,00 kr
    429,00 kr
    1 * 960 Items
       429,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2336
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    2 - 200 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 772,00 kr
    772,00 kr
    1 * 960 Items
       772,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2431
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    2 - 200 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 386,00 kr
    386,00 kr
    1 * 480 Items
       386,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2342
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    2 - 200 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 811,00 kr
    811,00 kr
    1 * 960 Items
       811,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2407
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
5 - 300 µl, length: 53 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked BIO-CERT® Liquid Handling Quality
    5 - 300 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 536,00 kr
    536,00 kr
    1 * 960 Items
       536,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732190
    Environmentally Preferable
    New Product
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    5 - 300 µl
    -
    1 pose, 1000
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 281,00 kr
    281,00 kr
    1 * 1.000 Items
       281,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2324
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk,CERTIFIED LIFE SCIENCE QUALITY
    5 - 300 µl
    -
    10 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10.000 Items
    1 * 10.000 Items - 2.530,00 kr
    2.530,00 kr
    1 * 10.000 Items
       2.530,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2331
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    5 - 300 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 429,00 kr
    429,00 kr
    1 * 960 Items
       429,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2337
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    5 - 300 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 772,00 kr
    772,00 kr
    1 * 960 Items
       772,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2432
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    5 - 300 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 386,00 kr
    386,00 kr
    1 * 480 Items
       386,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2343
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    5 - 300 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 811,00 kr
    811,00 kr
    1 * 960 Items
       811,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2408
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
50 - 1000 µl, length: 70 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked BIO-CERT® Liquid Handling Quality
    50 - 1000 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 547,00 kr
    547,00 kr
    1 * 960 Items
       547,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732192
    Environmentally Preferable
    New Product
  • Bulk, blue, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1000 µl
    -
    2 poser, 500 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 265,00 kr
    265,00 kr
    1 * 1.000 Items
       265,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2325
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, blue, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1000 µl
    -
    10 poser, 500 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5.000 Items
    1 * 5.000 Items - 1.070,00 kr
    1.070,00 kr
    1 * 5.000 Items
       1.070,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2332
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1000 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 437,00 kr
    437,00 kr
    1 * 960 Items
       437,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2338
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1000 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 791,00 kr
    791,00 kr
    1 * 960 Items
       791,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2433
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1000 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 397,00 kr
    397,00 kr
    1 * 480 Items
       397,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2344
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1000 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 906,00 kr
    906,00 kr
    1 * 960 Items
       906,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2409
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
1 - 50 µl, length: 50 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    1 - 50 µl
    -
    2 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2.000 Items
    1 * 2.000 Items - 518,00 kr
    518,00 kr
    1 * 2.000 Items
       518,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2322
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    1 - 50 µl
    -
    10 poser, 1000 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10.000 Items
    1 * 10.000 Items - 2.070,00 kr
    2.070,00 kr
    1 * 10.000 Items
       2.070,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2328
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    1 - 50 µl
    -
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 463,00 kr
    463,00 kr
    1 * 960 Items
       463,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2335
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    1 - 50 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 837,00 kr
    837,00 kr
    1 * 960 Items
       837,00 kr
    Specifikationer:
      Reduktion af affald|Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2430
    Environmentally Preferable
    Reduktion af affald|Lav fremstillingspåvirkning
      Reduktion af affald|Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    1 - 50 µl
    -
    5 Tip-Box, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 405,00 kr
    405,00 kr
    1 * 480 Items
       405,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2341
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    1 - 50 µl
    +
    10 racks, 96 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 881,00 kr
    881,00 kr
    1 * 960 Items
       881,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-2403
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
50 - 1250 µl, length 86 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1250 µl
    -
    -
    2 bags, 500 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 600,00 kr
    600,00 kr
    1 * 1.000 Items
       600,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7275
    Environmentally Preferable
  • Bulk, XXL, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1250 µl
    -
    -
    6 bags, 500 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 3.000 Items
    1 * 3.000 Items - 1.540,00 kr
    1.540,00 kr
    1 * 3.000 Items
       1.540,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7277
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    -
    5 racks , 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 797,00 kr
    797,00 kr
    1 * 480 Items
       797,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7171
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    -
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 980,00 kr
    980,00 kr
    1 * 480 Items
       980,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7173
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    -
    5 boxes , 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 997,00 kr
    997,00 kr
    1 * 480 Items
       997,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7175
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    -
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.400,00 kr
    1.400,00 kr
    1 * 480 Items
       1.400,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7163
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    +
    5 racks , 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.370,00 kr
    1.370,00 kr
    1 * 480 Items
       1.370,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3971
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    +
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.530,00 kr
    1.530,00 kr
    1 * 480 Items
       1.530,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3973
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    +
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.560,00 kr
    1.560,00 kr
    1 * 480 Items
       1.560,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3976
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    +
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.760,00 kr
    1.760,00 kr
    1 * 480 Items
       1.760,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3969
    Environmentally Preferable
50 - 1250 µl XL, filtered, length: 102 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    +
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.370,00 kr
    1.370,00 kr
    1 * 480 Items
       1.370,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3975
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    +
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.530,00 kr
    1.530,00 kr
    1 * 480 Items
       1.530,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3972
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    +
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.560,00 kr
    1.560,00 kr
    1 * 480 Items
       1.560,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3974
    Environmentally Preferable
  • TipBox, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    +
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.760,00 kr
    1.760,00 kr
    1 * 480 Items
       1.760,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    732-3970
    Environmentally Preferable
50 - 1250 µl XL, length: 102 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1250 µl
    -
    -
    2 bags, 500 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 600,00 kr
    600,00 kr
    1 * 1.000 Items
       600,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7276
    Environmentally Preferable
  • Bulk, XXL, CERTIFIED LIFE SCIENCE QUALITY
    50 - 1250 µl
    -
    -
    6 bags, 500 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 3.000 Items
    1 * 3.000 Items - 1.540,00 kr
    1.540,00 kr
    1 * 3.000 Items
       1.540,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7178
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    -
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 797,00 kr
    797,00 kr
    1 * 480 Items
       797,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7177
    Environmentally Preferable
  • TipRack, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    -
    5 racks, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 980,00 kr
    980,00 kr
    1 * 480 Items
       980,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7167
    Environmentally Preferable
  • TipBox, XL, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    -
    -
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 997,00 kr
    997,00 kr
    1 * 480 Items
       997,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7165
    Environmentally Preferable
  • TipBox, XL, BIO-CERT® LIQUID HANDLING QUALITY
    50 - 1250 µl
    +
    -
    5 boxes, 96 each
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 480 Items
    1 * 480 Items - 1.400,00 kr
    1.400,00 kr
    1 * 480 Items
       1.400,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    613-7172
    Environmentally Preferable
0,5 - 5 ml, length: 160 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    500 - 5000 µl
    -
    1 pose, 200
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 219,00 kr
    219,00 kr
    1 * 200 Items
       219,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    612-5754
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    500 - 5000 µl
    -
    5 poser, 200 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.010,00 kr
    1.010,00 kr
    1 * 1.000 Items
       1.010,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    612-5701
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, CERTIFIED LIFE SCIENCE QUALITY
    500 - 5000 µl
    -
    1 Tip-Box, 28 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 28 Items
    1 * 28 Items - 387,00 kr
    387,00 kr
    1 * 28 Items
       387,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3841
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
1 - 10 ml, length: 156,5 mm
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Bulk, CERTIFIED LIFE SCIENCE QUALITY
    1000 - 10000 µl
    -
    2 poser, 100 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 Items
    1 * 200 Items - 410,00 kr
    410,00 kr
    1 * 200 Items
       410,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3445
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • Bulk, XXL, CERTIFIED LIFE SCIENCE QUALITY
    1000 - 10000 µl
    -
    100 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 1.950,00 kr
    1.950,00 kr
    1 * 1.000 Items
       1.950,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3446
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
  • TipBox, CERTIFIED LIFE SCIENCE QUALITY
    1000 - 10000 µl
    -
    1 Tip-Box, 18 pr. stk.
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 18 Items
    1 * 18 Items - 500,00 kr
    500,00 kr
    1 * 18 Items
       500,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-3842
    Environmentally Preferable
    Lav fremstillingspåvirkning
      Lav fremstillingspåvirkning
Produktinformation
  • Spidstype
    Volumen
    Steril
    Filter
    Pakket
    Tilgængelighed
    Price
  • Racked BIO-CERT® Liquid Handling Quality
    1 - 50 µl
    -
    1 pack = 960 piece(s) (10 racks×96 each)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 960 Items
    1 * 960 Items - 579,00 kr
    579,00 kr
    1 * 960 Items
       579,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRND732186
    Environmentally Preferable
    New Product
Accessories
Produktinformation
  • Katalog #
    Farvekode
    Tilgængelighed
    Price
  • Empty TipBox, PP, with tip-tray, stackable, for tips up to 50 µl
    Empty TipBox, PP, with tip-tray, stackable, for tips up to 50 µl
    613-2424
    grå
    Log ind for tilgængelighed
    1 * 1 Items - 46,10 kr
Produktinformation
  • Katalog #
    Farvekode
    Tilgængelighed
    Price
  • Empty TipBox, PP, with tip-tray, stackable, for tips up to 1000 µl
    Empty TipBox, PP, with tip-tray, stackable, for tips up to 1000 µl
    613-2427
    blå
    Log ind for tilgængelighed
    1 * 1 Items - 53,90 kr
Produktinformation
  • Katalog #
    Farvekode
    Tilgængelighed
    Price
  • Empty TipBox, PP, with tip-tray, stackable, for tips up to 300 µl
    Empty TipBox, PP, with tip-tray, stackable, for tips up to 300 µl
    613-2426
    grøn
    Log ind for tilgængelighed
    1 * 1 Items - 53,90 kr
Produktinformation
  • Katalog #
    Farvekode
    Tilgængelighed
    Price
  • Empty TipBox, PP, with tip-tray, stackable, for tips up to 200 µl
    Empty TipBox, PP, with tip-tray, stackable, for tips up to 200 µl
    613-2425
    gul
    Log ind for tilgængelighed
    1 * 1 Items - 46,10 kr