Cut resistant gloves, KryTech 582
: MAPA
These seamless HDPE liners with nitrile GRIP & PROOF coating are liquidproof and provide a good grip of oily parts.
- Optimal cut resistance: level 5
- Excellent resistance to abrasion
- Visible indicator (blue textile) to easily identify the cut-performance level in the workplace
- Performances of the gloves guaranteed after 5 cleaning cycles in washing machine
- Non-slip grip
Ideal for metal working, the assembly of thin metal sheets, metal rolling and stamping, handling and sorting sharp parts, finishing work as well as for handling glass sheets.
: EN 388:2016 (4X43D), ISO 13997: 18N (1835g) (ANSI Cut Level 4); Cat. II.
These seamless HDPE liners with nitrile GRIP & PROOF coating are liquidproof and provide a good grip of oily parts.
- Optimal cut resistance: level 5
- Excellent resistance to abrasion
- Visible indicator (blue textile) to easily identify the cut-performance level in the workplace
- Performances of the gloves guaranteed after 5 cleaning cycles in washing machine
- Non-slip grip
Ideal for metal working, the assembly of thin metal sheets, metal rolling and stamping, handling and sorting sharp parts, finishing work as well as for handling glass sheets.
: EN 388:2016 (4X43D), ISO 13997: 18N (1835g) (ANSI Cut Level 4); Cat. II.
- Catalog No:
- MAPA34582037
- MAPA34582038
- MAPA34582039
- MAPA34582030
- MAPA34582031
- Model:
- KryTech 582
- KryTech 582
- KryTech 582
- KryTech 582
- KryTech 582
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- HDPE
- HDPE
- HDPE
- HDPE
- HDPE
- Overflade:
- 3/4 Grip & Proof coating
- 3/4 Grip & Proof coating
- 3/4 Grip & Proof coating
- 3/4 Grip & Proof coating
- 3/4 Grip & Proof coating
- Manchet:
- Strikket
- Strikket
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 388:2016 (4X43D)
- EN 388:2016 (4X43D)
- EN 388:2016 (4X43D)
- EN 388:2016 (4X43D)
- EN 388:2016 (4X43D)
Product Family Options
- Handske størrelseFarveLængde
Specifications
Cat. No.MAPA34582037ModelKryTech 582Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeHDPEOverflade3/4 Grip & Proof coatingManchetStrikketEN normEN 388:2016 (4X43D)Specifications
Cat. No.MAPA34582038ModelKryTech 582Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeHDPEOverflade3/4 Grip & Proof coatingManchetStrikketEN normEN 388:2016 (4X43D)Specifications
Cat. No.MAPA34582039ModelKryTech 582Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeHDPEOverflade3/4 Grip & Proof coatingManchetStrikketEN normEN 388:2016 (4X43D)Specifications
Cat. No.MAPA34582030ModelKryTech 582Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeHDPEOverflade3/4 Grip & Proof coatingManchetStrikketEN normEN 388:2016 (4X43D)Specifications
Cat. No.MAPA34582031ModelKryTech 582Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeHDPEOverflade3/4 Grip & Proof coatingManchetStrikketEN normEN 388:2016 (4X43D)