VWR®, Standard solution (1000 ppm Ca) for the preparation of calcium standard solution (100 ppm Ca), alcoholic Reag. Ph. Eur., 5000802
Catalog # 88085.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Koncentration:1000 mg/L
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Calcium
- Symbol:Ca
Specifications
About this item
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.