Capillary columns, TRACE™ TR-WaxMS
Kapillær, TR-WaxMS, 1,00 µm, 0,25 mm, 30 m
Katalog # 555-9087
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
TRACE™ TR-WaxMS GC columns are ideal for high-polarity separations as part of GC/MS analysis.
- Polar phase, polyethylene glycol
- Proprietary bonding method for expanded operating temperatures
- Extremely low bleed for improved sensitivity and library matches
- High stability with oxygen and water
Specifikationer
- Kolonne form:Kapillær
- Filmtykkelse:1,00 µm
- Ø indv.:0,25 mm
- Længde:30 m
- Fase:TR-WaxMS