Test tubes, round bottom, with GL screw cap
120 mm, 16 mm, GL 18, 1,8 mm
Katalog # KAVA632422631612
Leverandør: Kavalier
Gå til:
Product Details & Documents
Test tubes, round bottom, with GL screw cap.
Specifikationer
- Udv.Ø:16 mm
- Gevind:GL 18
- Vægtykkelse:1,8 mm
- Længde:120 mm