Order Entry
Denmark
ContactUsLinkComponent
D-(+)-Glucose-[¹³C₆,1,2,3,4,5,6,6-²H₇] (98% ¹³C , 50% D)
D-(+)-Glucose-[¹³C₆,1,2,3,4,5,6,6-²H₇] (98% ¹³C , 50% D)
Catalog # CAMBCDLM-3813-1
CAS Number:  
D-(+)-Glucose-[¹³C₆,1,2,3,4,5,6,6-²H₇] (98% ¹³C , 50% D)
Catalog # CAMBCDLM-3813-1
CAS Number:  

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:201417-01-8
Formula:C₆H₅D₇O₆
MDL Number:MFCD00274634