Orders LinkContactUsLinkComponent
Arachidonic acid
Arachidonic acid
Glass bottle for solids, 100 mg
Katalog # ROTH4717.1
Marketsource
Leverandør:   Roth Carl
Arachidonic acid
Katalog # ROTH4717.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  506-32-1
Arachidonic acid
Glass bottle for solids, 100 mg
CAS nummer:  506-32-1
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents
Synonymer:  (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
  • CAS nummer:506-32-1
  • Formel:C₂₀H₃₂O₂
  • MDL nummer:MFCD00004417
  • Molecular Weight:304.5 g/mol
  • EINECS:208-033-4
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 mg