Woulff bottles
Leverandør: witeg Labortechnik
Gå til:
Om denne vare
Made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion gives relatively high resistance to temperature changes
With two side necks GL 18, center neck GL 25, complete with screw caps.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion gives relatively high resistance to temperature changes
With two side necks GL 18, center neck GL 25, complete with screw caps.