Repetitive pipettes, mechanical, DISTRIMAN®
Catalog # 612-7810
Supplier: GILSON
Specifications
- Kapacitet:1 - 1250 µl
- Beskrivelse:Manual repetitive pipette, DISTRIMAN®
- Environmentally Preferable:
- Pakn:1
Specifications
About this item
DISTRIMAN® is a positive-displacement, continuously adjustable repeater-pipette to simplify multiple dispensing when used with Gilson DISTRITIP® syringes.
- Zero risk of accidental conversion errors with the direct read out
- Time saving: continuous adjustable volume and numerous distribution of aliquots (up to 125)
- Faster multiple dispensing
- Control button for volume selection
- DISTRITIP® syringes are available in standard and sterilised versions
DISTRIMAN® can dispense aliquots ranging in volume from 1 µl to 1,25 ml with the combination of its three dedicated models of DISTRITIP®. Each syringe can dispense up to 125 aliquots (of the minimal volume).
Ideal for any application requiring repetitive dispensing in the clinical, hospital, biological, chemical, food & beverage, pharmaceutical, cosmetics and forensics fields.