Orders LinkContactUsLinkComponent
Retort stand, Azlon®
Retort stand, Azlon®
Leverandør:   SciLabware
Retort stand, Azlon®
Leverandør:  SciLabware
Certifikater
Retort stand, Azlon®
Retort stand, Azlon®
2 tilgængelige muligheder nedenfor
Product Details & Documents
Weighted retort stand made from white PE.
  • Weighs 2,5 kg ensuring stability
  • Corrosion resistant
  • Polyethylene coated metal rod screws into the base
  • Available with the hole in the centre or at the short edge
Product Family Options
Produktinformation
  • Vægt
    B×D×H
    Armlængde
    Farve
    Tilgængelighed
    Price
  • 241-0001
    Retort stand, with hole at the centre, rod size 760×12,5 mm
    Retort stand, with hole at the centre, rod size 760×12,5 mm
    2,5 kg
    171×248×52 mm
    760 mm
    hvid
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.190,00 kr
    2.190,00 kr
    1 * 1 Items
       2.190,00 kr
  • 241-0002
    Retort stand, with hole at the edge, rod size 760×12,5 mm
    Retort stand, with hole at the edge, rod size 760×12,5 mm
    2,5 kg
    171×248×52 mm
    760 mm
    hvid
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.190,00 kr
    2.190,00 kr
    1 * 1 Items
       2.190,00 kr
This product has been blocked by your organization. Please contact your purchasing department for more information