4'-(1-Hydroxy-2-methylpropyl)acetophenone
Glass bottle for solids, 100 mg
Katalog # PROOMM0002.51
Leverandør: LGC Standards PROMOCHEM
4'-(1-Hydroxy-2-methylpropyl)acetophenone
Katalog # PROOMM0002.51
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
- Formel:C₁₂H₁₆O₂
- MDL nummer:MFCD08275598
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg