Hesperidin
Glass bottle for solids, 10 g
Katalog # ROTH7737.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
- CAS nummer:520-26-3
- Formel:C₂₈H₃₄O₁₅
- MDL nummer:MFCD00075663
- Molecular Weight:610.56 g/mol
- EINECS:208-288-1
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



