Membrane filters, hydrophobic edge
Supplier: Merck
Certificates
About this item
Hydrophobic edge membranes are standard MF-type filters with edges rendered hydrophobic to 3 or 6 mm widths. In antibiotic and bacteriostatic drug sterility testing by membrane filter method, the 6 mm wide non-wetting edge prevents drug intrusion under holder sealing rim.
- Available in sterile or autoclave as ready packs
Applications include microbiological and sterility testing of antibiotics and bacteriostatic drugs and filling machine operations.
Filter surface: plain
Filter color: white
Specifications
- Catalog No:
- MILXGSEP047S0
- MILXHAEG047S0
- MILXHAEG047AW
- MILFSMEP0470W
- Model:
- GSEP
- HAEG, gridded
- HAEG, gridded
- SMEP
- Porestørrelse:
- 0,22 µm
- 0,45 µm
- 0,45 µm
- 5,00 µm
- Tykkelse:
- -
- -
- -
- 135 µm
- Flowhastighed for vand:
- -
- -
- -
- 580 l/min/cm²
- Flowhastighed for luft:
- -
- -
- -
- 32 l/min/cm²
- Max. temperature:
- 75 °C