Drum funnels, PP
350 mm, 30 mm
Katalog # 221-1153
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Funnels, made of PP, for filling containers with large quantities of liquid or powdered substances quickly.
- Very good chemical resistance
- Very good temperature resistance
- Quick and easy to clean
Specifikationer
- Tragt udv. Ø:350 mm
- Stilk udv. Ø:30 mm