Orders LinkContactUsLinkComponent
Pressure filter holders
Pressure filter holders
Leverandør:   Merck
Pressure filter holders
Leverandør:  Merck
Certifikater
Pressure filter holders
2 tilgængelige muligheder nedenfor
Product Details & Documents
These filter holders are ideal for ultra-cleaning or sterilising batch volumes of liquids.
  • Filter holder has reservoir to hold liquid for batch filtration
  • Autoclavable with membrane in place for sterile filtration
The cylindrical barrel holds 100 or 340 ml of liquid, which is filtered by externally applied gas pressure through a suitable filter supported on the holder base.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • MILFXX4004700
  • MILFXX4004740
  • Filtreringsareal:
  • 11,3 cm²
  • 11,3 cm²
  • Indløb/udløb forbindelser:
  • 9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
  • 9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
  • Max. inlet pressure:
  • 6,9 bar
  • 6,9 bar
Specifikationer
Product Family Options
Produktinformation
  • Materiale
    Kapacitet
    Til Ø
    Højde
    Udv.Ø
    Tilgængelighed
    Price
  • MILFXX4004700
    Pressure filter holder
    Pressure filter holder
    Rustfrit stål
    100 ml
    47 mm
    292 mm
    70 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 15.820,00 kr
    15.820,00 kr
    1 * 1 Items
       15.820,00 kr
    Specifikationer:
    More
    Specifications
    Filtreringsareal
    11,3 cm²
    Cat. No.
    MILFXX4004700
    Indløb/udløb forbindelser
    9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
    Max. inlet pressure
    6,9 bar
  • MILFXX4004740
    Pressure filter holder
    Pressure filter holder
    Rustfrit stål
    340 ml
    47 mm
    533 mm
    70 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 19.740,00 kr
    19.740,00 kr
    1 * 1 Items
       19.740,00 kr
    Specifikationer:
    More
    Specifications
    Filtreringsareal
    11,3 cm²
    Cat. No.
    MILFXX4004740
    Indløb/udløb forbindelser
    9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
    Max. inlet pressure
    6,9 bar
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filter support screen, stainless steel, 47 mm
    Filter support screen, stainless steel, 47 mm
    MILFXX4004704
    Log ind for tilgængelighed
    1 * 1 Items - 1.890,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Locking ring gasket, PTFE
    Locking ring gasket, PTFE
    MILFXX4004714
    Log ind for tilgængelighed
    1 * 5 Items - 1.230,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement parts kit, 47 mm holder includes 5 each top cap O-rings, locking ring gasket, filter sealing O-ring and screen gasket
    Replacement parts kit, 47 mm holder includes 5 each top cap O-rings, locking ring gasket, filter sealing O-ring and screen gasket
    MILFXX40047RK
    Log ind for tilgængelighed
    1 * 1 Items - 3.700,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • PVC tubing 3 m with I.Ø. 9,5 mm
    PVC tubing 3 m with I.Ø. 9,5 mm
    MILFXX6700034
    Log ind for tilgængelighed
    1 * 1 Items - 816,00 kr