Cleaning trolley system, Origo
Om denne vare
The Origo trolley system has been designed for maximum flexibility and changeability. The user can either choose one of the standard configurations or have the trolley customised exactly to his requirements, and when requirements change so the system can be reconfigured. The 300 series offer the abitility to retro-fit and re-customise the trolley over and over, offering almost limitless possibilities.
- Adjustable handlebars
- Height adjustable trays and drawers
- Wide choice of castor size great manoeuvrability and thought-through design result in a state-of-the-art cleaning tool
Origo 100:
The Origo 100 is the entry model with a plastic base, fixed configuration and a fool-proof, no-tool assembly.
Origo 300:
The Origo 300 is the configurable alternative to the 100, again using a plastic base plate.
2 tilgængelige muligheder nedenfor
Product Details & Documents
The Origo trolley system has been designed for maximum flexibility and changeability. The user can either choose one of the standard configurations or have the trolley customised exactly to his requirements, and when requirements change so the system can be reconfigured. The 300 series offer the abitility to retro-fit and re-customise the trolley over and over, offering almost limitless possibilities.
- Adjustable handlebars
- Height adjustable trays and drawers
- Wide choice of castor size great manoeuvrability and thought-through design result in a state-of-the-art cleaning tool
Origo 100:
The Origo 100 is the entry model with a plastic base, fixed configuration and a fool-proof, no-tool assembly.
Origo 300:
The Origo 300 is the configurable alternative to the 100, again using a plastic base plate.
Product Family Options
Produktinformation
- TilgængelighedPrice
- 129-0850Cleaning trolley, Origo 100 FX:
- 1 base plate L
- 3 trays
- 4 buckets (2×6 l, 1×15 l and 1×25 l)
- 1 Ultraspeed press with G-lever
- 1 bin bag frame, plastic 120 l
- 2 clamping wire metal
- 1 bin bag lid, 120 l
- 2 displays for cleaning plan
- 1 extension
- 3 handle holders
- 1 frame holder
- castor size: 100 mmCleaning trolley, Origo 100 FX:
- 1 base plate L
- 3 trays
- 4 buckets (2×6 l, 1×15 l and 1×25 l)
- 1 Ultraspeed press with G-lever
- 1 bin bag frame, plastic 120 l
- 2 clamping wire metal
- 1 bin bag lid, 120 l
- 2 displays for cleaning plan
- 1 extension
- 3 handle holders
- 1 frame holder
- castor size: 100 mmLog ind for tilgængelighed1 * 1 Items4.930,00 kr - VILE121594Cleaning trolley, Origo 300 HX:
- 2 trays
- 2 buckets, 6 L
- 2 Mopbox
- 2 Mopbox lid
- 1 bin bag holder plastic 120 L with metal clamps
- 1 mop holder
- 1 handle holderCleaning trolley, Origo 300 HX:
- 2 trays
- 2 buckets, 6 L
- 2 Mopbox
- 2 Mopbox lid
- 1 bin bag holder plastic 120 L with metal clamps
- 1 mop holder
- 1 handle holderLog ind for tilgængelighed1 * 1 Items6.750,00 kr
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Pre-box to prepare mops or cloths, including colour coding clips (without lid)Pre-box to prepare mops or cloths, including colour coding clips (without lid)129-0707Log ind for tilgængelighed1 * 1 Items - 278,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Swep sieve; fits on top of prepmop box for dosing mops on or off trolley, pierced with holes to ensure even dosing of mopsSwep sieve; fits on top of prepmop box for dosing mops on or off trolley, pierced with holes to ensure even dosing of mops129-0709Log ind for tilgængelighed1 * 1 Items - 426,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Swep lid for pre mop box
Produktinformation
- Katalog #TilgængelighedPrice
- Bin bag support 70 L
Produktinformation
- Katalog #TilgængelighedPrice
- Lid for bin bag, 70 L with clips
Produktinformation
- Katalog #TilgængelighedPrice
- Lid for bucket Origo, 6 L
Produktinformation
- Katalog #TilgængelighedPrice
- Display for cleaning plan
Produktinformation
- Katalog #TilgængelighedPrice
- Tray for Origo cleaning system
Produktinformation
- Katalog #TilgængelighedPrice
- Half tray for Origo cleaning system
Produktinformation
- Katalog #TilgængelighedPrice
- Lid for bin bag, 120 L with clips
Produktinformation
- Katalog #TilgængelighedPrice
- Origo 500 HH - base plate
Produktinformation
- Katalog #TilgængelighedPrice
- Prebox origo box without color code
Produktinformation
- Katalog #TilgængelighedPrice
- Origo 300/500 bin bag frame metal 120 LOrigo 300/500 bin bag frame metal 120 LVILE122557Log ind for tilgængelighed1 * 1 Items - 843,00 kr