Labels for I.D. PRO® Plus, LS2000 and BradyMarker™ printer
BRDY032466
: Brady
- Model:B-619
- Farve:hvid
- Beskrivelse:Printer labels
- Dimensioner:38×31 mm
- Materiale:Polyester
- Seq:10
- Pakket:250 labels/roll
- Pakn:1 Roll
- Klæbestof:Permanent akryl