Orders LinkContactUsLinkComponent
Polyribonucleotide poly(I)·poly(C)
Polyribonucleotide poly(I)·poly(C)
Poly(I)·Poly(C), 265 mg
Katalog # 27-4732-01
Leverandør:   Cytiva
Polyribonucleotide poly(I)·poly(C)
Katalog # 27-4732-01
Leverandør:  Cytiva
CAS nummer:  
Polyribonucleotide poly(I)·poly(C)
Polyribonucleotide poly(I)·poly(C)
Poly(I)·Poly(C), 265 mg
CAS nummer:  
Price
19.720,00 kr
Log ind for tilgængelighed
1 * 265 mg
Product Details & Documents
The double-stranded polyribonucleotide poly(I)·poly(C) induces interferon in rabbit cells, and there has been interest in its effectiveness for inducing human interferon in vitro.
Specifikationer
Specifikationer
  • Pakn:
    265 mg
  • Beskrivelse:
    Poly(I)·Poly(C)