Orders LinkContactUsLinkComponent
Protein A/G HP SpinTrap™ Buffer Kit
Protein A/G HP SpinTrap™ Buffer Kit
Protein A/G HP SpinTrap™ Buffer kit, 1 KIT
Katalog # 28-9135-67
Leverandør:   Cytiva
Protein A/G HP SpinTrap™ Buffer Kit
Katalog # 28-9135-67
Leverandør:  Cytiva
CAS nummer:  
Protein A/G HP SpinTrap™ Buffer Kit
Protein A/G HP SpinTrap™ Buffer Kit
Protein A/G HP SpinTrap™ Buffer kit, 1 KIT
CAS nummer:  
Price
2.490,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Protein A/G HP SpinTrap™ Buffer kit is optimised for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with protein A HP SpinTrap™, protein G HP SpinTrap™, and Ab SpinTrap™ columns.
  • High quality buffers
  • 2.5% acetic acid elution buffer
  • All buffer components listed and defined
  • Convenient format
  • Stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment
The buffers provided in the kit are optimized for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with Protein A HP SpinTrap, Protein G HP SpinTrap, and Ab SpinTrap columns. Kit includes 10x Binding/Washing Buffer [TBS]: 0.5 M Tris, 1.5 M NaCl, pH 7.5 Elution Buffer [Classic]: 2.5 % HAc, 10x Elution Buffer [Crosslink]: 1 M glycine-HCl, pH 2.9, 10x Crosslink Solution A: 2 M triethanolamine, pH 8.9, 10x Crosslink Solution B: 1 M ethanolamine, pH 8.9.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Protein A/G HP SpinTrap™ Buffer kit
Specifikationer