Orders LinkContactUsLinkComponent
Emergency rescue sling, Miller®
Emergency rescue sling, Miller®
2×4 m
Catalog # 111-1389
Supplier:   HONEYWELL SAFETY
Emergency rescue sling, Miller®
Catalog # 111-1389
Supplier:  HONEYWELL SAFETY
Emergency rescue sling, Miller®
Emergency rescue sling, Miller®
2×4 m
Price
4.120,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Used by many leading rescue services around the world, this comfortable and easy to use rescue sling is based on a helicopter sling: The chest strap can be undone and fitted around the casualty while remaining attached to the rescue system.
  • Adjustable leg straps allow any angle of casualty lift
  • Cell foam padding (9 mm) on the chest strap enhances comfort for the casualty
  • Two additional attachment rings regulate the size for smaller casualties
Certifications: EN 1497
Specifications
  • Længde:
    2×4 m