Orders LinkContactUsLinkComponent
Chemical Resistant Overalls, 4515
Chemical Resistant Overalls, 4515
 :   3M
Chemical Resistant Overalls, 4515
 :  3M
Chemical Resistant Overalls, 4515
Chemical Resistant Overalls, 4515
These disposable overalls made of highly breathable SMS material provide basic protection against dry particles, certain liquid chemical splashes and hazardous dusts such as asbestos fibres.
  • Breathable material technology for reduced heat stress
  • Robust, low linting
  • Two-way zip with storm flap (incl. tape tabs) for convenience and additional protection
  • Elasticated cuffs, waist and ankles for a better fit and freedom of movement
Typical light duty applications may include asbestos inspection, insulation laying, cement manufacturing and metal grinding and polishing.
Certifications: Type 5/6, EN 1073-2; Cat. III
  • Catalog No:
  • 113-6259
  • 113-6258
  • 113-6257
  • 113-6260
  • 113-6254
  • 113-6255
  • 113-6256
  • 113-6265
  • 113-6264
  • 113-6263
  • 113-6266
  • 113-6261
  • 113-6262
  • 113-6279
  • 113-6278
  • 113-6277
  • 113-6280
  • MMMA4515WXXL
  • 113-6275
  • 113-6276
  • Model:
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • 4515
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Outfit applikationer:
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
Product Family Options
  • Farve
    Tøj størrelse
  • blå
    S
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6259
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    M
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6258
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    L
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6257
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    XL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6260
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    XXL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6254
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    3XL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6255
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • blå
    4XL
    1 * 20 Items
    1 * 20 Items - 1.190,00 kr
    1.190,00 kr
    1 * 20 Items
       1.190,00 kr
    Specifications
    Cat. No.
    113-6256
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    S
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6265
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    M
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6264
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    L
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6263
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    XL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6266
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    XXL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6261
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • orange
    3XL
    1 * 20 Items
    1 * 20 Items - 1.170,00 kr
    1.170,00 kr
    1 * 20 Items
       1.170,00 kr
    Specifications
    Cat. No.
    113-6262
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    S
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6279
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    M
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6278
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    L
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6277
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    XL
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6280
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    XXL
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    MMMA4515WXXL
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    3XL
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6275
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial
  • hvid
    4XL
    1 * 20 Items
    1 * 20 Items - 1.060,00 kr
    1.060,00 kr
    1 * 20 Items
       1.060,00 kr
    Specifications
    Cat. No.
    113-6276
    Model
    4515
    Sleeve or leg length
    Long
    Outfit applikationer
    Chemical protection|Laboratory|Industrial