Orders LinkContactUsLinkComponent
Monitor, UVis-920
Monitor, UVis-920
Monitor UVis-920
Katalog # 11-0007-54
Leverandør:   Cytiva
Monitor, UVis-920
Katalog # 11-0007-54
Leverandør:  Cytiva
Monitor, UVis-920
Monitor, UVis-920
Monitor UVis-920
Price
61.330,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The monitor UVis-920 is a high-precision, multipurpose, standalone UV monitor. Use of different filters allows the monitor to cover wavelengths from 200 to 700 nm, without changing the lamp.
  • High precision UV/Vis absorbance detection over a wide flow range
  • Easy to use, change of wavelength by filter push-button mechanism without changing the lamp
  • Minimised band broadening and maximised signal-to-noise ratio
  • Minimised footprint
Specifikationer
  • Beskrivelse:
    Monitor UVis-920
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • UV flow cell, 10 mm, with optical fiber
    UV flow cell, 10 mm, with optical fiber
    18-1111-11
    Log ind for tilgængelighed
    1 * 1 Items - 19.100,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • UV flow cell, 2 mm, with optical fiber
    UV flow cell, 2 mm, with optical fiber
    18-1111-10
    Log ind for tilgængelighed
    1 * 1 Items - 18.840,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • UV flow cell ÄKTApilot, 1/2/5 mm, with optical fiber
    UV flow cell ÄKTApilot, 1/2/5 mm, with optical fiber
    11-0008-50
    Log ind for tilgængelighed
    1 * 1 Items - 22.460,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filter with filter holder, 260 nm
    Filter with filter holder, 260 nm
    11-0007-34
    Log ind for tilgængelighed
    1 * 1 Items - 14.200,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filter with filter holder, 280 nm
    Filter with filter holder, 280 nm
    11-0007-35
    Log ind for tilgængelighed
    1 * 1 Items - 3.830,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Banana pin connector, black
    Banana pin connector, black
    18-1004-24
    Log ind for tilgængelighed
    1 * 1 Items - 340,00 kr