Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Con A 4B
Affinity chromatography columns, HiTrap™ Con A 4B
Leverandør:   Cytiva
Affinity chromatography columns, HiTrap™ Con A 4B
Leverandør:  Cytiva
Certifikater
Affinity chromatography columns, HiTrap™ Con A 4B
Affinity chromatography columns, HiTrap™ Con A 4B
2 tilgængelige muligheder nedenfor
Product Details & Documents
HiTrap™ Con A 4B is a ready-to-use column prepacked with Con A Sepharose™ 4B, a medium for convenient separation and purification of glycoproteins, polysaccharides, and glycolipids.
  • Con A Sepharose™ is concanavalin A coupled to Sepharose™ 4B
  • Can be operated with a syringe, a peristaltic pump, or a liquid chromatography system such as ÄKTA design
Concanavalin A (Con A) is a tetrameric metalloprotein isolated from Canavalia ensiformis (jack bean). Con A binds molecules containing α-D-mannopyranosyl, α-D-glucopyranosyl, and sterically related residues. The binding sugar requires the presence of C-3, C-4 and C-5 hydroxyl groups for reaction with Con A.
HiTrap™ Con A 4B columns can be operated with a syringe, a peristaltic pump, or a liquid chromatography system such as ÄKTA design.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 28-9520-85
  • 28-9520-96
  • Maks. tryk:
  • 5 bar
  • 5 bar
  • pH-område:
  • 4 - 9
  • 4 - 9
Specifikationer
Product Family Options
Produktinformation
  • Kolonnemateriale
    Partikelstørrelse
    Ø indv.
    Længde
    Kapacitet
    Flowhastighed
    Tilgængelighed
    Price
  • 28-9520-85
    HiTrap™ Con A 4B
    HiTrap™ Con A 4B
    PP
    90 µm
    7,00 mm
    25 mm
    5×1 ml
    <4 ml/min
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 Items
    1 * 5 Items - 1.640,00 kr
    1.640,00 kr
    1 * 5 Items
       1.640,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    28-9520-85
    Maks. tryk
    5 bar
    pH-område
    4 - 9
  • 28-9520-96
    HiTrap™ Con A 4B
    HiTrap™ Con A 4B
    PP
    90 µm
    16,00 mm
    25 mm
    5×5 ml
    <20 ml/min
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 Items
    1 * 5 Items - 6.170,00 kr
    6.170,00 kr
    1 * 5 Items
       6.170,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    28-9520-96
    Maks. tryk
    5 bar
    pH-område
    4 - 9