Orders LinkContactUsLinkComponent
HPLC columns, VariTide RPC
HPLC columns, VariTide RPC
Supplier:   VARIAN
HPLC columns, VariTide RPC
Supplier:  VARIAN
Certificates
HPLC columns, VariTide RPC
5 Options Available Below
Product Details & Documents
VariTide RPC column is designed for use across the pH range, making it suitable for the analysis and purification of acid or basic and hydrophilic or hydrophobic peptides, using acid, neutral and basic eluents.
  • Separations run at elevated temperature and pH, and under acidic, neutral and basic conditions
  • Robustness and durability
  • Single, easy to use, universal column
  • A single column to cover the full range of synthetic peptides
A high efficiency, small particle packing, with a pore size optimised for high capacity across the peptide size range, from 5 to 60 amino acids.
Product Family Options
Varitide RPC columns
Product Information
  • Beskrivelse
    Partikelstørrelse
    Ø indv.
    Længde
    Availability
    Price
  • Columns, VariTide, for synthetic peptides
    6 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Columns, VariTide, for synthetic peptides
    6 µm
    10,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Columns, VariTide, for synthetic peptides
    6 µm
    21,20 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
VariTide RPC bulk media
Product Information
  • Beskrivelse
    Partikelstørrelse
    Ø indv.
    Længde
    Availability
    Price
  • VariTide RPC bulk media, 100 g
    6 µm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • VariTide RPC bulk media, 1 kg
    6 µm