Orders LinkContactUsLinkComponent
First-Strand cDNA Synthesis Kit
First-Strand cDNA Synthesis Kit, 55 Tests
Katalog # 27-9261-01
Leverandør:   Cytiva
First-Strand cDNA Synthesis Kit
Katalog # 27-9261-01
Leverandør:  Cytiva
CAS nummer:  
First-Strand cDNA Synthesis Kit
First-Strand cDNA Synthesis Kit
First-Strand cDNA Synthesis Kit, 55 Tests
CAS nummer:  
Price
4.090,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Designed to generate full length first-strand cDNA from mRNA templates. Following first-strand cDNA synthesis, the sample can be used directly for in vitro amplification using PCR or for second-strand synthesis using the Gubler-Hoffman method.
  • For synthesis of single-stranded cDNA prior to amplification by PCR
  • Features preassembled bulk reaction mixes to save time and minimise potential for pipetting errors
  • Allows users to choose primers
Includes a choice of two primers: pd(N)6 random hexamers and NotI-(dT)18.
 : Kit includes sufficient for up to 55 first-strand syntheses, each of which will produce enough material for up to 15 amplification reactions: Bulk First-Strand cDNA Reaction Mixes, mRNA Standard, DTT Solution, RNase-Free Water, pd(N)6 Primer, NotI-(dT)18 Bifunctional Primer and instruction booklet.
Specifikationer
Specifikationer
  • Pakn:
    55 Tests
  • Beskrivelse:
    First-Strand cDNA Synthesis Kit