PIG® vertical mat roll holder
Vertical mat roll holder, 510×510×1120 mm
Katalog # NEWPGEN249
Leverandør: NEW PIG
Gå til:
Product Details & Documents
No more lugging around large rolls of mat, let this vertical mat roll holder do all the work.
- No-tip swivel wheel casters allow holder to move freely in any direction
- Constructed of powder-coated, hot-rolled steel with a comfortable handle
- 8 cm high platform is low enough for easy mat positioning
For use with any PIG mat roll up to 93 cm width x 51 cm diameter, holds up to 41 kg.
Specifikationer
- Beskrivelse:Vertical mat roll holder
- B×D×H:510×510×1120 mm