Orders LinkContactUsLinkComponent
VWR®, Storage boxes for sample vials/bottles
rød, 130×130×52 mm
Katalog # 548-1487A
Leverandør:   VWR Collection
VWR®, Storage boxes for sample vials/bottles
Katalog # 548-1487A
Leverandør:  VWR Collection
VWR®, Storage boxes for sample vials/bottles
VWR®, Storage boxes for sample vials/bottles
rød, 130×130×52 mm
Price
1.600,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Storage boxes for convenient, safe storage and transportation of sample vials and sample bottles. A selection of boxes to securely hold a wide range of vial sizes. Ideal for space-saving storage in fridges, as the transparent lid prevents condensation on the closures so avoiding contamination in the cooling unit.
  • Temperature resistant from −80 to 100 °C
  • Alphanumeric coding (1,5 and 4 ml) for clear sample identification
  • Chemically resistant, fairly robust and stackable
  • PP, coloured
Specifikationer
  • Farve:
    rød
  • Seq:
    02
  • B×D×H:
    130×130×52 mm