Streptavidin, Cy3®
1 mg
PA43001
: Cytiva
Streptavidin-fluor conjugates suitable for use in the fluorescent analysis of biotinylated samples.
- Streptavidin conjugated to a range of fluorescent dyes for use in in situ hybridisation and flow cytometry
- High specificity reagents
This product is supplied as a lyophilized solid in phosphate-buffered saline (0.02 M potassium phosphate, 0.15 M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide. Reconstitute with 1.0 ml of deionized water, dispense into suitable aliquots and freeze. Avoid repeated freeze-thaw cycles.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
- Pakn:1 mg
- Cat. No.:PA43001
- Konjugation:Cy3®
- Form:Solid
- Protein/peptide name:Streptavidin
- pH:7.2
- Tested applications:Suitable for use in the fluorescent analysis of biotinylated samples
- Farve:Blue