0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Flasker
Plastflasker
Bottles, wide neck, with screw cap
Bottles, wide neck, with screw cap
SEMA0128
:
SEMADENI
Bottles, wide neck, with screw cap
SEMA0128
:
SEMADENI
Kapacitet:
5000 ml
B×D×H:
190×180×230 mm
Hals indv. Ø:
109 mm
Pakn:
1
HDPE, translucent.
Good chemical resistance
Good temperature resistance