Orders LinkContactUsLinkComponent
Bottles, wide neck, LDPE, rectangular, with screw cap
Bottles, wide neck, LDPE, rectangular, with screw cap
5000 ml, 63 mm, 180×140×290 mm
Katalog # SEMA0065
Leverandør:   SEMADENI
Bottles, wide neck, LDPE, rectangular, with screw cap
Katalog # SEMA0065
Leverandør:  SEMADENI
Bottles, wide neck, LDPE, rectangular, with screw cap
Bottles, wide neck, LDPE, rectangular, with screw cap
5000 ml, 63 mm, 180×140×290 mm
Price
684,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
LDPE, translucent.
  • Good chemical resistance
  • Godd temperature resistance
Specifikationer
  • Kapacitet:
    5000 ml
  • B×D×H:
    180×140×290 mm
  • Hals indv. Ø:
    63 mm