Orders LinkContactUsLinkComponent
Pep-Extract™
Pep-Extract™
Pep-Extract™, 100 Tests
Katalog # 786-243
Leverandør:   G-Biosciences
Pep-Extract™
Katalog # 786-243
Leverandør:  G-Biosciences
CAS nummer:  
Pep-Extract™
Pep-Extract™, 100 Tests
CAS nummer:  
Price
1.570,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Following digestion of proteins with proteases, the addition of Pep-Extract™ rapidly elutes digested peptides in under 20 minutes. The eluted peptides can be used directly in mass spectrometry analysis without any additional clean up. Contains sufficient for 500 protein spots.
Specifikationer
Specifikationer
  • Pakn:
    100 Tests
  • Beskrivelse:
    Pep-Extract™