Orders LinkContactUsLinkComponent
SilverOUT™, G-Biosciences
SilverOUT™, G-Biosciences
SilverOUT™, 100 Tests
Catalog # 786-244
Supplier:   G-Biosciences
SilverOUT™, G-Biosciences
Catalog # 786-244
Supplier:  G-Biosciences
CAS Number:  
SilverOUT™, G-Biosciences
SilverOUT™, G-Biosciences
SilverOUT™, 100 Tests
CAS Number:  
Price
1.570,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
SilverOUT removes interfering silver ions from gel pieces.
Gel spots can be destained within 10 minutes.
Compatible with downstream processes including mass spectrometry.
Documents
Specifications
Specifications
  • Pakn:
    100 Tests
  • Beskrivelse:
    SilverOUT™